Merck
  • The active zone protein family ELKS supports Ca2+ influx at nerve terminals of inhibitory hippocampal neurons.
The active zone protein family ELKS supports Ca2+ influx at nerve terminals of inhibitory hippocampal neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-09-12)
Changliang Liu, Lydia S Bickford, Richard G Held, Hajnalka Nyitrai, Thomas C Südhof, Pascal S Kaeser
ABSTRACT
In a presynaptic nerve terminal, synaptic vesicle exocytosis is restricted to specialized sites called active zones. At these sites, neurotransmitter release is determined by the number of releasable vesicles and their probability of release. Proteins at the active zone set these parameters by controlling the presynaptic Ca(2+) signal, and through docking and priming of synaptic vesicles. Vertebrate ELKS proteins are enriched at presynaptic active zones, but their functions are not well understood. ELKS proteins are produced by two genes in vertebrates, and each gene contributes ∼50% to total brain ELKS. We generated knock-out mice for ELKS1 and found that its constitutive removal causes lethality. To bypass lethality, and to circumvent redundancy between ELKS1 and ELKS2 in synaptic transmission, we used a conditional genetic approach to remove both genes in cultured hippocampal neurons after synapses are established. Simultaneous removal of ELKS1 and ELKS2 resulted in a 50% decrease of neurotransmitter release at inhibitory synapses, paralleled by a reduction in release probability. Removal of ELKS did not affect synapse numbers or their electron microscopic appearance. Using Ca(2+) imaging, we found that loss of ELKS caused a 30% reduction in single action potential-triggered Ca(2+) influx in inhibitory nerve terminals, consistent with the deficits in synaptic transmission and release probability. Unlike deletion of the active zone proteins RIM, RIM-BP, or bruchpilot, ELKS removal did not lead to a measurable reduction in presynaptic Ca(2+) channel levels. Our results reveal that ELKS is required for normal Ca(2+) influx at nerve terminals of inhibitory hippocampal neurons.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-ELKS antibody, Mouse monoclonal, clone ELKS-30, purified from hybridoma cell culture