Merck
Immobilized rennin in TC/SG composite in cheese production.
Food chemistry (2016-02-03)
Eleftheria Barouni, Theano Petsi, Dionysios Kolliopoulos, Dimitrios Vasileiou, Panagiotis Panas, Argyro Bekatorou, Maria Kanellaki, Athanasios A Koutinas
ABSTRACT
The object of the current study was to develop a new process for continuous Feta-type cheese production using a biocatalyst consisting of immobilized rennin on a tubular cellulose/starch gel (TC/SG) composite, which has been proven to be an appropriate carrier for enzyme immobilization. Different methodologies were used in order to prepare four biocatalysts. The most effective was selected for cheese production in a 1L continuous system, providing two economically useful results for the dairy industries: (i) increase of productivity by the continuous coagulation of milk, and (ii) saving of the rennin enzyme expenses of the batch coagulation of milk. The criteria used to choose the appropriate biocatalyst was based on the time of coagulation in successive batches, the concentration of immobilized rennin combined with the filter efficiency and its application in the continuous system. Physicochemical analyses of the cheeses at various stages of the ripening were performed. No significant differences compared to cheeses prepared with the traditional method were found. Aroma compounds were determined by SPME GC-MS.
Product Description
Millipore
M-17 Agar, suitable for microbiology, NutriSelect® Basic
Millipore
Malt Extract Agar, suitable for microbiology, NutriSelect® Plus
Millipore
Violet Red Bile Agar, suitable for microbiology, NutriSelect® Plus
Millipore
Violet Red Bile Glucose Agar, suitable for microbiology, NutriSelect® Plus
Millipore
MRS Agar, suitable for microbiology, NutriSelect® Basic