Merck
Nitrated fatty acids: synthesis and measurement.
Free radical biology & medicine (2012-12-04)
Steven R Woodcock, Gustavo Bonacci, Stacy L Gelhaus, Francisco J Schopfer
ABSTRACT
Nitrated fatty acids are the product of nitrogen dioxide reaction with unsaturated fatty acids. The discovery of peroxynitrite and peroxidase-induced nitration of biomolecules led to the initial reports of endogenous nitrated fatty acids. These species increase during ischemia/reperfusion, but concentrations are often at or near the limits of detection. Here, we describe multiple methods for nitrated fatty acid synthesis and sample extraction from complex biological matrices and a rigorous method of qualitative and quantitative detection of nitrated fatty acids by liquid chromatography-mass spectrometry. In addition, optimized instrument conditions and caveats regarding data interpretation are discussed.
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl alcohol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Pricing and availability is not currently available.
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Pricing and availability is not currently available.