Merck
  • Diffusion and retention are major determinants of protein targeting to the inner nuclear membrane.
Diffusion and retention are major determinants of protein targeting to the inner nuclear membrane.
The Journal of cell biology (2015-06-10)
Rosemarie Ungricht, Michael Klann, Peter Horvath, Ulrike Kutay
ABSTRACT
Newly synthesized membrane proteins are constantly sorted from the endoplasmic reticulum (ER) to various membranous compartments. How proteins specifically enrich at the inner nuclear membrane (INM) is not well understood. We have established a visual in vitro assay to measure kinetics and investigate requirements of protein targeting to the INM. Using human LBR, SUN2, and LAP2β as model substrates, we show that INM targeting is energy-dependent but distinct from import of soluble cargo. Accumulation of proteins at the INM relies on both a highly interconnected ER network, which is affected by energy depletion, and an efficient immobilization step at the INM. Nucleoporin depletions suggest that translocation through nuclear pore complexes (NPCs) is rate-limiting and restricted by the central NPC scaffold. Our experimental data combined with mathematical modeling support a diffusion-retention-based mechanism of INM targeting. We experimentally confirmed the sufficiency of diffusion and retention using an artificial reporter lacking natural sorting signals that recapitulates the energy dependence of the process in vivo.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Pricing and availability is not currently available.