Merck
  • Interleukin-17 Stimulates STAT3-Mediated Endothelial Cell Activation for Neutrophil Recruitment.
Interleukin-17 Stimulates STAT3-Mediated Endothelial Cell Activation for Neutrophil Recruitment.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-08-19)
Shaopeng Yuan, Shimeng Zhang, Yuanyuan Zhuang, He Zhang, Jinye Bai, Qi Hou
ABSTRACT
Interleukin-17 (IL-17) is a major pro-inflammatory cytokine that initiates and maintains inflammation. However, the molecular mechanisms as to how IL-17 influences endothelial cells to promote neutrophil recruitment are not fully understood. Human endothelial cells (HMECs) were stimulated with IL-17, and investigated for proliferation, migration, and tubule formation activities. Transwell chemotaxis and adhesion assays were performed to assess neutrophil recruitment. Cytokine production was measured by Cytokine Array Chip and ELISA. Western blotting and immunofluorescent analysis were used to detect the phosphorylation and translocation of STAT3. Specific inhibitors, small interfering RNA, and phosphorylation mutants were used to confirm that IL-17 induced STAT3 activation via IL-17RA signaling. Activation of HMECs with IL-17 induced STAT3 phosphorylation and nuclear translocation, which were associated with induction of GRO-α, GM-CSF and IL-8, and neutrophil recruitment. Phosphorylation of STAT3 was identified mainly at the tyrosine in position 705 (Y705), and the Y705F mutants attenuated IL-17-mediated STAT3 activation. Moreover, specific inhibitors, FLLL31, or siRNA silencing of STAT3 attenuated HMECs activation, resulting in inhibition of GRO-α, GM-CSF, IL-8 production, and neutrophil recruitment. Furthermore, phosphorylation of STAT3 was identified as downstream of IL-17RA signaling. IL-17 induced STAT3 activation as a necessary step in endothelial cell activation and neutrophil recruitment.
Sigma-Aldrich
Interleukin-17A human, IL-17A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3
Sigma-Aldrich
FLLL31, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
Interleukin-17A human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O