Merck
  • Crimean-Congo haemorrhagic fever replication interplays with regulation mechanisms of apoptosis.
Crimean-Congo haemorrhagic fever replication interplays with regulation mechanisms of apoptosis.
The Journal of general virology (2014-12-08)
Helen Karlberg, Yee-Joo Tan, Ali Mirazimi
ABSTRACT
Pathogenesis of viral haemorrhagic fevers is associated with alteration of vascular barrier function and haemorrhage. To date, the specific mechanism behind this is unknown. Programmed cell death and regulation of apoptosis in response to viral infection is an important factor for host or virus survival but this has not been well-studied in the case of Crimean-Congo hemorrhagic fever virus (CCHFV). In this study, we demonstrated that CCHFV infection suppresses cleavage of poly(ADP-ribose) polymerase (PARP), triggered by staurosporine early post-infection. We also demonstrated that CCHFV infection suppresses activation of caspase-3 and caspase-9. Most interestingly, we found that CCHFV N can suppress induction of apoptosis by Bax and inhibit the release of cytochrome c from the inner membrane of mitochondria to cytosol. However, CCHFV infection induces activation of Bid late post-infection, suggesting activation of extrinsic apoptotic signalling. Consistently, supernatant from cells stimulated late post-infection was found to induce PARP cleavage, most probably through the TNF-α death receptor pathway. In summary, we found that CCHFV has strategies to interplay with apoptosis pathways and thereby regulate caspase cascades. We suggest that CCHFV suppresses caspase activation at early stages of the CCHFV replication cycle, which perhaps benefits the establishment of infection. Furthermore, we suggest that the host cellular response at late stages post-infection induces host cellular pro-apoptotic molecules through the death receptor pathway.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, FCC, FG