Merck
  • Effect of chemical interaction on the bonding strengths of self-etching adhesives to deproteinised dentine.
Effect of chemical interaction on the bonding strengths of self-etching adhesives to deproteinised dentine.
Journal of dentistry (2015-06-10)
Liqun Zhou, Yake Wang, HongYe Yang, Jinxin Guo, Franklin R Tay, Cui Huang
ABSTRACT
The present study examined (1) the chemical interaction between three self-etching adhesives and sodium hypochlorite (NaOCl)-deproteinised dentine, and (2) the influence of NaOCl treatment on bond strength of self-etching adhesives with/without adhesive functional monomers to dentine. Caries-free dentine disks (control) and those treated with 5.25% NaOCl for 60s were prepared. Xeno V (no functional monomers), G-Bond (containing 4-MET) or S3 Bond (containing 10-MDP) were applied to the NaOCl-treated dentine and either left without further treatment, or rinsed with 100% ethanol or distilled water. Attenuated total reflection (ATR) spectroscopy and field-emission scanning electron microscopy (FE-SEM) were used to evaluate the affinity of functional monomers with deproteinised dentine. Chemical interaction between the functional monomers and deproteinised dentine was evaluated using thin-film X-ray diffraction (TF-XRD). Microtensile bond strength (MTBS) was used to evaluate the mechanical property of the adhesives, either immediately or after thermo-cycling (5-55°C) for 10,000 cycles. According to the ATR and FE-SEM results, G-Bond and S3 Bond showed stronger affinity to deproteinised dentine than Xeno V even after rinsing with water. TF-XRD showed that chemical interaction between S3 Bond and deproteinised dentine occurred by formation of 10-MDP-Ca salt. Both deproteinisation and thermo-cycling adversely affected the MTBS of Xeno V (P<0.05) but deproteinisation had no significant influence on S3 Bond. When bonding to NaOCl-treated dentine, self-etch adhesives containing functional monomers (10-MDP) can maintain immediate and aged bond strengths after 10,000 thermal cycles.
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Germanium, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, rod, 6mm, diameter 6.0mm, single crystal, 100%
Germanium, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%
Germanium, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
Germanium, sheet, 6x6mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 10x10mm, thickness 0.5mm, single crystal, -111, 100%
Germanium, sheet, 7x24mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, sheet, 10x10mm, thickness 0.6mm, single crystal, -111, 100%
Germanium, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
Germanium, sheet, 25x25mm, thickness 1.0mm, single crystal, 99.999%
Germanium, sheet, 50x50mm, thickness 0.5mm, single crystal, 99.999%
Germanium, sheet, 25x25mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%