Merck
  • Effective Two-Photon Excited Photodynamic Therapy of Xenograft Tumors Sensitized by Water-Soluble Bis(arylidene)cycloalkanone Photosensitizers.
Effective Two-Photon Excited Photodynamic Therapy of Xenograft Tumors Sensitized by Water-Soluble Bis(arylidene)cycloalkanone Photosensitizers.
Journal of medicinal chemistry (2015-09-24)
Qianli Zou, Hongyou Zhao, Yuxia Zhao, Yanyan Fang, Defu Chen, Jie Ren, Xiaopu Wang, Ying Wang, Ying Gu, Feipeng Wu
ABSTRACT
A series of bis(arylidene)cycloalkanone photosensitizers modified by polyethylene glycol (PEG) have been studied for two-photon excited photodynamic therapy (2PE-PDT). As compared with their prototype compounds, these PEGylated photosensitizers show enhanced water solubilities while their photophysical and photochemical properties, including linear absorption, two-photon absorption, fluorescence, and singlet oxygen quantum yield, remain unaltered. In vitro behaviors (cellular uptake, subcellular localization, photocytotoxicity in both PDT and 2PE-PDT) of these photosensitizers reveal that an optimized lipid-water partition coefficient can be obtained by adjusting the length and position of the PEG chains. Among them, the photosensitizer modified asymmetrically by two tetraethylene glycol chains presents the best performance as a 2PE-PDT candidate. Selective blood-vessel closure and obvious therapeutic effect in inhibiting the growth of tumors are confirmed by in vivo 2PE-PDT after intravenous injection of this photosensitiezer. The survival periods of treated tumor-bearing mice are significantly prolonged. This study demonstrates the feasibility of using a simple molecule to construct a potential candidate for 2PE-PDT.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
4-Diethylaminobenzaldehyde, 99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
1,3-Diphenylisobenzofuran, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis