Merck
  • Improvement in fingerprint detection using Tb(III)-dipicolinic acid complex doped nanobeads and time resolved imaging.
Improvement in fingerprint detection using Tb(III)-dipicolinic acid complex doped nanobeads and time resolved imaging.
Forensic science international (2015-06-06)
Frank M Hauser, Gerd Knupp, Simon Officer
ABSTRACT
This paper deals with the synthesis and application of lanthanide complex doped nanobeads used as a luminescent fingerprint powder. Due to their special optical properties, namely a long emission lifetime, sharp emission profiles and large Stokes shifts, luminescent lanthanide complexes are useful for discriminating against signals from background emissions. This is a big advantage because latent fingerprints placed on multicoloured fluorescent surfaces are difficult to develop with conventional powders. The complex of 2,6-dipicolinic acid (DPA) and terbium ([Tb(DPA)3](3-)) is used for this purpose. Using the Stöber process, this complex is incorporated into a silica matrix forming nanosized beads (230-630nm). It is shown that the [Tb(DPA)3](3-) is successfully incorporated into the beads and that these beads exhibit the wanted optical properties of the complex. A phenyl functionalisation is applied to increase the lipophilicity of the beads and finally the beads are used to develop latent fingerprints. A device for time resolved imaging was built to improve the contrast between developed fingerprint and different background signals, whilst still detecting the long lasting luminescence of the complex. The developed fingerprint powder is therefore promising to develop fingerprints on multicoloured fluorescent surfaces.
Product Description
Terbium, powder, 1g, max. particle size 500 micron, 99.9%
Terbium, powder, 5g, max. particle size 500 micron, 99.9%
Terbium, powder, 2g, max. particle size 500 micron, 99.9%
Terbium, rod, 100mm, diameter 6.35mm, cast, 99%
Terbium, rod, 50mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
D-Lactose monohydrate, SAJ special grade
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
Triethoxyphenylsilane, 98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Nitric acid, SAJ first grade, 65.0-66.0%, density: 1.40