Merck
  • Improved radiosynthesis and preliminary in vivo evaluation of a (18)F-labeled glycopeptide-peptoid hybrid for PET imaging of neurotensin receptor 2.
Improved radiosynthesis and preliminary in vivo evaluation of a (18)F-labeled glycopeptide-peptoid hybrid for PET imaging of neurotensin receptor 2.
Bioorganic & medicinal chemistry (2015-02-19)
Simone Maschauer, Cornelia Greff, Jürgen Einsiedel, Julian Ott, Philipp Tripal, Harald Hübner, Peter Gmeiner, Olaf Prante
ABSTRACT
The neurotensin receptor 2 (NTS2) is an attractive target for cancer imaging, as it is overexpressed in a variety of tumor types including prostate, pancreas and breast carcinoma. The aim of this study was the development of the first NTS2 subtype selective (18)F-labeled radioligand for imaging NTS2 expression in vivo by positron emission tomography (PET). The radiosynthesis of glycopeptoid (18)F-4 was realized by copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC), applying the prosthetic group 6-deoxy-6-[(18)F]fluoroglucosyl azide for (18)F-fluoroglycosylation of the alkyne-terminated NT(8-13) analog Pra-N-Me-Arg-Arg-Pro-N-homo-Tyr-Ile-Leu-OH. The binding affinity of the peptide-peptoid 4 for NTS2 was 7nM with excellent subtype selectivity over NTS1 (260-fold). In vitro autoradiography studies of rat brain slices confirmed the high selectivity of (18)F-4 for NTS2. Biodistribution experiments using HT29 and PC3 tumor-bearing nude mice revealed high renal and only moderate tumor uptake, while PET imaging experiments revealed specific binding of (18)F-4 in NTS2-positive tumors. As (18)F-4 displayed high stability in vitro but fast degradation in vivo, future work will focus on the development of metabolically more stable NT(8-13) analogs.
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, amber vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 1000
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, amber vial, pack of 100
Whatman® Mini-UniPrep® G2 slit septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, clear vial, pack of 100
Whatman® Mini-UniPrep® G2 slit septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.45 μm, clear vial, pack of 100
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 1000
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)
Tantalum(V) ethoxide, packaged for use in deposition systems
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 1000
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 100
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 100
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 1000