Merck
  • Jak3, STAT3, and STAT5 inhibit expression of miR-22, a novel tumor suppressor microRNA, in cutaneous T-Cell lymphoma.
Jak3, STAT3, and STAT5 inhibit expression of miR-22, a novel tumor suppressor microRNA, in cutaneous T-Cell lymphoma.
Oncotarget (2015-08-06)
Nina A Sibbesen, Katharina L Kopp, Ivan V Litvinov, Lars Jønson, Andreas Willerslev-Olsen, Simon Fredholm, David L Petersen, Claudia Nastasi, Thorbjørn Krejsgaard, Lise M Lindahl, Robert Gniadecki, Nigel P Mongan, Denis Sasseville, Mariusz A Wasik, Lars Iversen, Charlotte M Bonefeld, Carsten Geisler, Anders Woetmann, Niels Odum
ABSTRACT
Aberrant activation of Janus kinase-3 (Jak3) and its key down-stream effectors, Signal Transducer and Activator of Transcription-3 (STAT3) and STAT5, is a key feature of malignant transformation in cutaneous T-cell lymphoma (CTCL). However, it remains only partially understood how Jak3/STAT activation promotes lymphomagenesis. Recently, non-coding microRNAs (miRNAs) have been implicated in the pathogenesis of this malignancy. Here, we show that (i) malignant T cells display a decreased expression of a tumor suppressor miRNA, miR-22, when compared to non-malignant T cells, (ii) STAT5 binds the promoter of the miR-22 host gene, and (iii) inhibition of Jak3, STAT3, and STAT5 triggers increased expression of pri-miR-22 and miR-22. Curcumin, a nutrient with anti-Jak3 activity and histone deacetylase inhibitors (HDACi) also trigger increased expression of pri-miR-22 and miR-22. Transfection of malignant T cells with recombinant miR-22 inhibits the expression of validated miR-22 targets including NCoA1, a transcriptional co-activator in others cancers, as well as HDAC6, MAX, MYCBP, PTEN, and CDK2, which have all been implicated in CTCL pathogenesis. In conclusion, we provide the first evidence that de-regulated Jak3/STAT3/STAT5 signalling in CTCL cells represses the expression of the gene encoding miR-22, a novel tumor suppressor miRNA.
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human JAK3
Sigma-Aldrich
MISSION® esiRNA, targeting human INSL3
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
3-Deazaneplanocin A hydrochloride, ≥97% (HPLC)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Sodium butyrate, Vetec, reagent grade, 99%