Merck
  • Confocal laser scanning microscopy elucidation of the micromorphology of the leaf cuticle and analysis of its chemical composition.
Confocal laser scanning microscopy elucidation of the micromorphology of the leaf cuticle and analysis of its chemical composition.
Protoplasma (2015-02-26)
Pavani P Nadiminti, James E Rookes, Ben J Boyd, David M Cahill
ABSTRACT
Electron microscopy techniques such as transmission electron microscopy (TEM) and scanning electron microscopy (SEM) have been invaluable tools for the study of the micromorphology of plant cuticles. However, for electron microscopy, the preparation techniques required may invariably introduce artefacts in cuticle preservation. Further, there are a limited number of methods available for quantifying the image data obtained through electron microscopy. Therefore, in this study, optical microscopy techniques were coupled with staining procedures and, along with SEM were used to qualitatively and quantitatively assess the ultrastructure of plant leaf cuticles. Leaf cryosections of Triticum aestivum (wheat), Zea mays (maize), and Lupinus angustifolius (lupin) were stained with either fat-soluble azo stain Sudan IV or fluorescent, diarylmethane Auramine O and were observed under confocal laser scanning microscope (CLSM). For all the plant species tested, the cuticle on the leaf surfaces could be clearly resolved in many cases into cuticular proper (CP), external cuticular layer (ECL), and internal cuticular layer (ICL). Novel image data analysis procedures for quantifying the epicuticular wax micromorphology were developed, and epicuticular waxes of L. angustifolius were described here for the first time. Together, application of a multifaceted approach involving the use of a range of techniques to study the plant cuticle has led to a better understanding of cuticular structure and provides new insights into leaf surface architecture.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Triacontane, 98%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%