Merck
  • Remediation of Hydrocarbon-Contaminated Soil by Washing with Novel Chemically Modified Humic Substances.
Remediation of Hydrocarbon-Contaminated Soil by Washing with Novel Chemically Modified Humic Substances.
Journal of environmental quality (2015-12-08)
César García-Díaz, Antonio Nebbioso, Alessandro Piccolo, Josefina Barrera-Cortés, Rafael Martínez-Palou
ABSTRACT
In this work, humic (HA) and fulvic acid (FA) were chemically modified by esterification and etherification with alkanes under microwave (MW) irradiation to improve their surfactant properties for the remediation of total petroleum hydrocarbons (TPHs)-contaminated soil. Humic acid and FA were evaluated as surfactant for the remediation of soil by means of washing an aged highly TPH-contaminated soil (50,000 mg TPH kg) sampled from a Mexican petrochemical area. The efficiency of chemical modification of HA and FA was increased and accelerated under MW irradiation with respect to that of conventional heating. Results showed that modified HA and FA were able to considerably reduce the contamination of TPH-polluted soils. The best results were obtained with HA modified by esterification with -dodecanol and FA modified with -decanol, which increased the hydrocarbon removal by 24 and 18%, respectively, with respect to amounts removed by the unmodified derivatives.
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Octanol, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
1-Bromopentane, 98%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Octanol, SAJ first grade, ≥75.0%
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
1-Decanol, analytical standard
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Bromotetradecane, 97%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%