Merck
Study of Cytotoxic Effects of Benzonitrile Pesticides.
BioMed research international (2015-09-05)
Petra Lovecka, Marketa Thimova, Petra Grznarova, Jan Lipov, Zdenek Knejzlik, Hana Stiborova, Tjokorda Gde Tirta Nindhia, Katerina Demnerova, Tomas Ruml
ABSTRACT
The benzonitrile herbicides bromoxynil, chloroxynil, dichlobenil, and ioxynil have been used actively worldwide to control weeds in agriculture since 1970s. Even though dichlobenil is prohibited in EU since 2008, studies addressing the fate of benzonitrile herbicides in the environment show that some metabolites of these herbicides are very persistent. We tested the cytotoxic effects of benzonitrile herbicides and their microbial metabolites using two human cell lines, Hep G2 and HEK293T, representing liver and kidneys as potential target organs in humans. The cell viability and proliferation were determined by MTT test and RTCA DP Analyzer system, respectively. The latter allows real-time monitoring of the effect of added substances. As the cytotoxic compounds could compromise cell membrane integrity, the lactate dehydrogenase test was performed as well. We observed high toxic effects of bromoxynil, chloroxynil, and ioxynil on both tested cell lines. In contrast, we determined only low inhibition of cell growth in presence of dichlobenil and microbial metabolites originating from the tested herbicides.
Sigma-Aldrich
2,6-Dichlorobenzonitrile, 97%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
2,6-Dichlorobenzoic acid, 98%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzamide, purified by sublimation, ≥99.5%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzamide, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)