Merck
  • Comparing indocyanine green, technetium, and blue dye for sentinel lymph node mapping in endometrial cancer.
Comparing indocyanine green, technetium, and blue dye for sentinel lymph node mapping in endometrial cancer.
Gynecologic oncology (2015-04-15)
J How, W H Gotlieb, J Z Press, J Abitbol, M Pelmus, A Ferenczy, S Probst, R Gotlieb, S Brin, S Lau
ABSTRACT
With the debate over extent of lymphadenectomy in endometrial cancer, sentinel lymph node (SLN) mapping may provide a focused approach to evaluate the most relevant lymph nodes (LN) while minimizing the complications. We evaluated SLN mapping using filtered technetium(99), indocyanine green (ICG), and blue dye. Prospective evaluation of 100 patients who underwent SLN mapping by using submucosal and deep stromal cervical injections of technetium(99), ICG, and blue dye as part of the staging for endometrial cancer. 286 SLNs were mapped (2.9 per patient) in 92% of patients. The bilateral detection rate was 76%. ICG had a significantly higher SLN detection rate than blue dye in both overall (87% vs 71%, respectively; p=0.005) and bilateral (65% vs 43%, respectively; p=0.002) detection, but similar SLN detection rates compared to technetium(99) in both overall (87% vs 88%, respectively; p=0.83) and bilateral (65% vs 71%, respectively; p=0.36) detection. In eight cases, the SLN was in the para-aortic area and in 14 cases in the pre-sacral, hypogastric vein, or parametrial area. In nine cases, the SLN was positive for metastasis, and in seven cases the SLN was the only positive node. One SLN was falsely negative. No complications or anaphylactic reactions occurred. Intra-operative SLN mapping using cervical injection is feasible in patients with endometrial cancer and yields adequate detection rates. It allows mapping of SLNs in areas (pre-sacral, hypogastric vein, parametrial) not routinely sampled. Given the poorer performance of blue dye, surgeons may omit its use if a combination of ICG and technetium(99) is used.
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Cardiogreen, polymethine dye