Merck
  • Diorganotin (IV) complexes with 4-nitro-N-phthaloyl-glycine: Synthesis, characterization, antitumor activity and DNA-binding studies.
Diorganotin (IV) complexes with 4-nitro-N-phthaloyl-glycine: Synthesis, characterization, antitumor activity and DNA-binding studies.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-05-12)
Chaoqun Yan, Jiali Zhang, Taigang Liang, Qingshan Li
ABSTRACT
Two novel diorganotin (IV) complexes, based on 4-nitro-N-phthaloyl-glycine (HL), namely {4-NO2C6H3(CO)2NCH2COO}2Sn(n-Bu)2 (1) and {4-NO2C6H3(CO)2NCH2COO}2SnMe2 (2), were synthesized and characterized by elemental analysis, FT-IR, (1)H- and (13)C-NMR spectroscopic techniques. In vitro antitumor activities of both complexes were evaluated by the 3-(4,5-dimethylthiazoly-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay against three human cancer cell lines: HepG-2 (human liver carcinoma), SGC-7901 (human gastric carcinoma) and LS174T (human colon carcinoma). Complex 1 exhibited strong antitumor activity with IC50 values of 1.51±0.41, 1.80±0.63, and 2.48±0.96 μM, respectively; while complex 2 had no obvious effects on the three selected cancer cell lines at high concentrations up to 100 μM. Complex 1-induced apoptosis was further confirmed by morphological observations and annexin V-FITC/PI staining flow cytometry analysis in HepG-2 cells. Cell cycle analysis revealed that complex 1 caused cell cycle arrest at G2/M phase. Molecular mechanism studies suggested that the apoptosis was mediated through the mitochondrial pathway with intracellular reactive oxygen species (ROS) promotion and mitochondrial membrane potential (MMP) disruption by finally activating effector caspase-3/9 to trigger cell apoptosis. Moreover, the interactions of both complexes with calf thymus DNA (CT-DNA) were investigated by using UV-Vis titration and fluorometric competition measurements. The DNA-binding constants Kb (intrinsic binding constant) and K(sv) (quenching constant) had been obtained in the order: 1>2, consisted with the antitumor activity results. Taken together, complex 1 exhibited excellent antitumor activity suggesting that it may be a potential candidate for further chemical optimization and cancer therapy.
Sigma-Aldrich
Dibutyltin dichloride, 96%
Sigma-Aldrich
Dimethyltin dichloride, 97%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis