Merck
  • Synthesis and Properties of a Novel Pyridineoxazoline Containing Optically Active Helical Polymer as a Catalyst Ligand for Asymmetric Diels-Alder Reaction.
Synthesis and Properties of a Novel Pyridineoxazoline Containing Optically Active Helical Polymer as a Catalyst Ligand for Asymmetric Diels-Alder Reaction.
Chirality (2015-04-25)
Heng Wang, Na Li, Jie Zhang, Xinhua Wan
ABSTRACT
A novel pyridineoxazoline (PyOx) containing helical polymer, poly{(-)-(S)-4-tert-butyl-2-[5-(4-tert-butylphenyl)-3-vinylpyridin-2-yl]-oxazoline} (PA), was designed and synthesized to approach the effect of chain conformation on the catalytic property. Its complex with Cu(OTf)(2) , i.e., Cu(II)-PA, was employed to catalyze the homogeneous Diels-Alder (D-A) reaction of alkenoyl pyridine N-oxides with cyclopentadiene in tetrahydrofuran. Compared with the previously reported copper complex, Cu(II)-P1 (RSC Advances, 2015, 5, 2882), which was derived from a nonhelical poly[(-)-(S)-4-tert-butyl-2-(3-vinylpyridin-2-yl)-oxazoline], Cu(II)-PA exhibited a remarkably enhanced enantioselectivity and reaction rate. However, its enantioselectivity was lower than the Cu(II) complex of (-)-(S)-4-tert-butyl-2-[5-(4-tert-butylphenyl)-3-vinylpyridin-2-yl]-oxazoline (Cu(II)-A), a low molar mass model compound.
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
4-tert-Butylphenylboronic acid, ≥95.0%
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, SAJ first grade, ≥99.0%
Sigma-Aldrich
N-Bromosuccinimide, ReagentPlus®, 99%
Sigma-Aldrich
Copper(II) trifluoromethanesulfonate, 98%
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
L-tert-Leucinol, 98%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder