Merck
  • Properties of short double-stranded RNAs carrying randomized base pairs: toward better controls for RNAi experiments.
Properties of short double-stranded RNAs carrying randomized base pairs: toward better controls for RNAi experiments.
RNA (New York, N.Y.) (2015-10-31)
Julian A Zagalak, Mirjam Menzi, Fabian Schmich, Hartmut Jahns, Afzal M Dogar, Florian Wullschleger, Harry Towbin, Jonathan Hall
ABSTRACT
Short interfering RNAs (siRNAs) are mediators of RNA interference (RNAi), a commonly used technique for selective down-regulation of target gene expression. Using an equimolar mixture of A, G, C, and U phosphoramidites during solid-phase synthesis, we introduced degenerate positions into RNA guide and passenger strands so that, when annealed, a large pool of distinct siRNA duplexes with randomized base pairs at defined sites was created. We assessed the randomization efficiency by deep sequencing one of the RNAs. All possible individual sequences were present in the pool with generally an excellent distribution of bases. Melting temperature analyses suggested that pools of randomized guide and passenger strands RNAs with up to eight degenerate positions annealed so that mismatched base-pairing was minimized. Transfections of randomized siRNAs (rnd-siRNAs) into cells led to inhibition of luciferase reporters by a miRNA-like mechanism when the seed regions of rnd-siRNA guide strands were devoid of degenerate positions. Furthermore, the mRNA levels of a select set of genes associated with siRNA off-target effects were measured and indicated that rnd-siRNAs with degenerate positions in the seed likely show typical non-sequence-specific effects, but not miRNA-like off-target effects. In the wake of recent reports showing the preponderance of miRNA-like off-target effects of siRNAs, our findings are of value for the design of a novel class of easily prepared and universally applicable negative siRNA controls.
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethoxytrimethylsilane, 98%
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%