Merck
  • Detecting the presence of hippocampus membrane androgen receptors in male SAMP8 mice and their induced synaptic plasticity.
Detecting the presence of hippocampus membrane androgen receptors in male SAMP8 mice and their induced synaptic plasticity.
Molecular and cellular endocrinology (2015-07-15)
Sha Li, Lin Kang, Yizhou Zhang, Baofeng Feng, Juan Du, Huixian Cui
ABSTRACT
Testosterone (T), the principal androgen, and its metabolite, dihydrotestosterone (DHT), are known to mediate their effects through binding to intracellular androgen receptors (iARs). In addition to their well-known genomic effects, androgens rapidly alter neuronal excitability through a non-genomic pathway mediated by membrane androgen receptors (mARs). The existence and specificity of mARs in the hippocampus were investigated in SAMP8 mice. Using T-BSA-FITC, we detected plasma membrane labeling by flow cytometry analysis for the presence of mARs. The specificity of binding was examined with iAR antagonist or anti-iAR antibody. Flow cytometry analysis showed that pretreatment with iAR antagonist, flutamide (F), failed to completely prevent the coupling action of the T-BSA-FITC membrane binding. In addition, we found classical iARs did not localize to the membrane of hippocampal neurons. These data indicate that these mARs might be not identical to classical iARs. Modulation of hippocampal synaptic plasticity by androgen has been attracting much attention. To identify the functional consequences induced by mARs, we analyzed the rapid effects of T on the density of dendritic spines using Golgi staining. The application of 50 μg/5 μl T and 30 μg/5 μl DHT induced a rapid increase in the dendritic spines within 2 h. Almost no difference was observed between T and T-BSA in the effect on thorn density. Next, we explored the protective mechanism and found that T and DHT altered the expression of synaptophysin (SYN) and postsynaptic dense material 95 (PSD95), which play crucial roles in cognitive function and synaptic plasticity.
Sigma-Aldrich
Testosterone 3-(O-carboxymethyl)oxime: BSA
Sigma-Aldrich
Testosterone 3-(O-carboxymethyl)oxime: BSA - fluorescein isothiocyanate conjugate
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%