Merck
  • Design of Zeolitic Imidazolate Framework Derived Nitrogen-Doped Nanoporous Carbons Containing Metal Species for Carbon Dioxide Fixation Reactions.
Design of Zeolitic Imidazolate Framework Derived Nitrogen-Doped Nanoporous Carbons Containing Metal Species for Carbon Dioxide Fixation Reactions.
ChemSusChem (2015-09-24)
Takashi Toyao, Mika Fujiwaki, Kenta Miyahara, Tae-Ho Kim, Yu Horiuchi, Masaya Matsuoka
ABSTRACT
Various N-doped nanoporous carbons containing metal species were prepared by direct thermal conversion of zeolitic imidazolate frameworks (ZIFs; ZIF-7, -8, -9, and -67) at different temperatures (600, 800, and 1000 °C). These materials were utilized as bifunctional acid-base catalysts to promote the reaction of CO2 with epoxides to form cyclic carbonates under 0.6 MPa of CO2 at 80 °C. The catalyst generated by thermal conversion of ZIF-9 at 600 °C (C600-ZIF-9) was found to exhibit a higher catalytic activity than the other ZIFs, other conventional catalysts, and other metal-organic framework catalysts. The results of various characterization techniques including elemental analysis, X-ray diffraction, X-ray photoelectron spectroscopy, X-ray absorption spectroscopy, and transmission electron microscopy show that C600-ZIF-9 contains partly oxidized Co nanoparticles and N species. Temperature-programmed desorption measurements by using CO2 and NH3 as probe molecules revealed that C600-ZIF-9 has both Lewis acid and Lewis base catalytic sites. Finally, the substrate scope was extended to seven other kinds of epoxides.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
tert-Butylbenzene, 99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Glycidyl isopropyl ether, 98%
Sigma-Aldrich
2-Methylimidazole, 99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)