Merck
  • Cleavable Molecular Beacon for Hg(2+) Detection Based on Phosphorothioate RNA Modifications.
Cleavable Molecular Beacon for Hg(2+) Detection Based on Phosphorothioate RNA Modifications.
Analytical chemistry (2015-06-11)
Po-Jung Jimmy Huang, Feng Wang, Juewen Liu
ABSTRACT
Mercury is a highly toxic heavy metal, and detection of Hg(2+) by biosensors has attracted extensive research interest in the past decade. In particular, a number of DNA-based sensing strategies have been developed. Well-known examples include thymine-Hg(2+) interactions and Hg(2+)-activated DNAzymes. However, these mechanisms are highly dependent on buffer conditions or require hybridization with another DNA strand. Herein, we report a new mechanism based on Hg(2+)-induced cleavage of phosphorothioate (PS) modified RNA. Among the various metal ions tested, Hg(2+) induced the most significant cleavage (∼16%), while other metals cleaved less than 2% of the same substrate. The uncleaved substrate undergoes desulfurization in the presence of Hg(2+). This cleavage reaction yields a similar amount of product from pH 3.5 to 7 and in the temperature range between 20 and 90 °C. Various PS RNA junctions can be cleaved with a similar efficiency, but PS DNA junctions cannot be cleaved. A molecular beacon containing three PS RNA modifications is designed, detecting Hg(2+) down to 1.7 nM with excellent selectivity. This sensor can also detect Hg(2+) in the Lake Ontario water sample, although its response is significantly masked by fish tissues.
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, suitable for plant cell culture