Merck
  • MiR-34a suppresses ovarian cancer proliferation and motility by targeting AXL.
MiR-34a suppresses ovarian cancer proliferation and motility by targeting AXL.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-04-22)
Rui Li, Xuejun Shi, Fengyu Ling, Chunguang Wang, Junxia Liu, Wei Wang, Ming Li
ABSTRACT
Increasing evidence has suggested that dysregulation of microRNAs (miRNAs) could contribute to tumor progression. The miR-34 family is directly transactivated by tumor suppressor p53 which is frequently mutated in various cancers; however, the effect of miR-34a on the ovarian cancer cells remains unclear. The aim of the paper was to study the expression of miR-34a in ovarian cancer and miR-34a's relation to the cell proliferation and metastasis in ovarian cancer in vitro. miR-34a expression was determined by quantitative RT-PCR in a panel of 60 human ovarian cancer samples. Functional characterization of miR-34a was accomplished by reconstitution of miR-34a expression in ovarian cancer cells by determining changes in proliferation, migration, and invasion. Our results showed that miR-34a is downregulated in ovarian cancer tissues compared with the corresponding adjacent non-neoplastic tissues, and the expression level of miR-34a was significantly lower in ovarian cancer cell lines in comparison with normal human fallopian tube epithelial cell line. The 3-(4,5)-dimethylthiahiazo(-z-y1)-3,5-diphenytetrazoliumromide (MTT) assay revealed significant cell proliferation inhibition in miR-34a transfectant compared with the control from HO8910 and SKOV3 cells, which displayed lowest expressions of miR-34a. Furthermore, the transwell assay also showed significant cell migration inhibition in miR-34a transfectant, compared with cell lines transfected with NC. Overexpression of miR-34a led to the inhibition of AXL expression, indicating that AXL is a target gene for miR-34a. Our data suggest that miR-34a may function as a tumor suppressor through repression of oncogenic AXL in ovarian cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human AXL
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Axl
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O