Merck
  • Peri-ovulatory putrescine supplementation reduces embryo resorption in older mice.
Peri-ovulatory putrescine supplementation reduces embryo resorption in older mice.
Human reproduction (Oxford, England) (2015-06-18)
Yong Tao, Dandan Liu, Guolong Mo, Hongmei Wang, X Johné Liu
ABSTRACT
Does peri-ovulatory putrescine supplementation of older mice improve oocyte quality and reduce the incidence of embryo resorption? Peri-ovulatory putrescine supplementation in older mice improved oocyte quality, as indicated by increased blastocyst cell numbers and reduced the incidence of embryo resorption. Rodents exhibit a transient rise of ornithine decarboxylase (ODC) and putrescine in the ovaries during ovulation. Older mice exhibit reduced ovarian ODC activity during ovulation. Supplementation of in vitro maturation medium with putrescine reduces oocyte aneuploidy rates of older mice. The rationale was to correct ovarian putrescine deficiency in older mice by peri-ovulatory putrescine supplementation in drinking water and to observe the reproductive consequences of this intervention. This project was conducted between 2010 and 2014. Older mice (9-11 months of age) were given regular drinking water (control) or drinking water with 1% putrescine dihydrochloride (62 mM) for 2-4 days before mating. Plugged mice were then withdrawn from putrescine supplementation. Blastocysts were retrieved on 3.5 days post coitum (dpc) for the determination of cell numbers. For resorption analyses, mice were killed on 9.5 dp or 12.5 dpc, and implantation sites were dissected to determine the embryo status. For birth studies, mice were examined every morning between 16.5 and 23.5 dpc. Births were recorded as live or stillbirth. We demonstrated that deficiency of ovarian putrescine in older mice can be restored by peri-ovulatory putrescine supplementation in drinking water. Putrescine supplementation in older mice increased blastocyst cell numbers (from 40 to 54; P < 0.0001, t-test), reduced embryo resorption rates (from 41.1 to 15.4% in old C57BL/6 mice, P < 0.0001, Fisher's exact test; from 14.2 to 6.4% in old CF1 mice, P = 0.004, Fisher's exact test), and doubled the number of live born pups. Furthermore, exogenous putrescine exhibited rapid absorption and excretion, and showed no toxicity to mothers or fetuses. The mechanism of putrescine action in oocytes and/or ovaries remains unclear. Peri-ovulatory putrescine deficiency in older mice appears to adversely impact on oocyte maturation resulting in poor quality embryos (as assessed by blastocyst cell numbers) and early embryo death. This study demonstrates a natural and simple remedy to improve oocyte quality in older women. This study was supported by the NSERC, the March of Dimes Foundation, and the National Natural Science Foundation of China. The authors declare no competing interest.
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Putrescine dihydrochloride, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Putrescine dihydrochloride, ≥98% (TLC)
Sigma-Aldrich
Putrescine dihydrochloride, powder, BioReagent, suitable for cell culture