Merck
  • Simultaneous Determination of Methanol, Ethanol and Formic Acid in Serum and Urine by Headspace GC-FID.
Simultaneous Determination of Methanol, Ethanol and Formic Acid in Serum and Urine by Headspace GC-FID.
Journal of analytical toxicology (2015-07-17)
Miroslava Bursová, Tomáš Hložek, Radomír Čabala
ABSTRACT
A simple, cost-effective headspace gas chromatography (GC) method coupled with GC with flame ionization detection for simultaneous determination of methanol, ethanol and formic acid was developed and validated for clinical and toxicological purposes. Formic acid was derivatized with an excess of isopropanol under acidic conditions to its volatile isopropyl ester while methanol and ethanol remained unchanged. The entire sample preparation procedure is complete within 6 min. The design of the experiment (the face-centered central composite design) was used for finding the optimal conditions for derivatization, headspace sampling and chromatographic separation. The calibration dependences of the method were quadratic in the range from 50 to 5,000 mg/L, with adequate accuracy (89.0-114.4%) and precision (<12%) in the serum. The new method was successfully used for determination of selected analytes in serum samples of intoxicated patients from among those affected by massive methanol poisonings in the Czech Republic in 2012.
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
N,N′-Bis(acryloyl)cystamine, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
3-Methyl-2-pentanone, 99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%