Merck
  • Snail and serpinA1 promote tumor progression and predict prognosis in colorectal cancer.
Snail and serpinA1 promote tumor progression and predict prognosis in colorectal cancer.
Oncotarget (2015-05-28)
Chae Hwa Kwon, Hye Ji Park, Jin Hwa Choi, Ja Rang Lee, Hye Kyung Kim, Hong-Jae Jo, Hyun Sung Kim, Nahmgun Oh, Geun Am Song, Do Youn Park
ABSTRACT
The role of Snail and serpin peptidase inhibitor clade A member 1 (serpinA1) in tumorigenesis has been previously identified. However, the exact role and mechanism of these proteins in progression of colorectal cancer (CRC) are controversial. In this study, we investigated the role of Snail and serpinA1 in colorectal cancer (CRC) and examined the mechanisms through which these proteins mediate CRC progression. Immunohistochemical analysis of 528 samples from patients with CRC showed that elevated expression of Snail or serpinA1 was correlated with advanced stage, lymph node metastasis, and poor prognosis. Moreover, we detected a correlation between Snail and serpinA1 expression. Functional studies performed using the CRC cell lines DLD-1 and SW-480 showed that overexpression of Snail or serpinA1 significantly increased CRC cell invasion and migration. Conversely, knockdown of Snail or serpinA1 expression suppressed CRC cell invasion and migration. ChIP analysis revealed that Snail regulated serpinA1 by binding to its promoter. In addition, fibronectin mediated Snail and serpinA1 signaling was involved in CRC cell invasion and migration. Taken together, our data showed that Snail and serpinA1 promoted CRC progression through fibronectin. These findings suggested that Snail and serpinA1 were novel prognostic biomarkers and candidate therapeutic targets in CRC.
Sigma-Aldrich
Anti-SERPINA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Anti-FN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution