Merck
  • The roles of ADAM33, ADAM28, IL-13 and IL-4 in the development of lung injuries in children with lethal non-pandemic acute infectious pneumonia.
The roles of ADAM33, ADAM28, IL-13 and IL-4 in the development of lung injuries in children with lethal non-pandemic acute infectious pneumonia.
Journal of clinical virology : the official publication of the Pan American Society for Clinical Virology (2014-12-03)
Emanuele Baurakiades, Victor Horácio Costa, Sonia Mara Raboni, Vivian Rafaela Telli de Almeida, Kelly Susana Kunze Larsen, Juliana Nemetz Kohler, Priscilla do Carmo Gozzo, Giseli Klassen, Graciele C M Manica, Lucia de Noronha
ABSTRACT
ADAM28, ADAM33, IL-13, IL-4 and other cytokines (IL-6 and IL-10) seem to play important roles in the persistence and maintenance of acute inflammatory processes that ultimately lead to lung remodeling and pulmonary fibrosis, which may be responsible for the high morbidity and mortality rates associated with non-pandemic acute viral pneumonias in childhood. The aim of this study was to evaluate the roles of ADAM33, ADAM28, IL4, IL6, IL10 and IL13 in the development of inflammation and alveolar fibrosis due to lethal acute respiratory infections of the lower airway in a pediatric population, especially in those with viral etiology. For this study, 193 cases were selected, and samples from the cases were processed for viral antigen detection by immunohistochemistry and then separated into two groups: virus-positive (n=68) and virus-negative (n=125). Immunohistochemistry was performed to assess the presence of metalloproteinases (ADAM33 and ADAM28) and inflammatory cytokines (IL-4, IL-13, IL-6, IL-10) in the alveolar septa. The virus-positive group showed stronger immunolabeling for ADAM33, ADAM28, IL-4 and IL-13 (p<0.0001 for all variables). The staining intensities for ADAM33 and ADAM28 were directly proportional to the intensities for IL-4 and IL-13 (p<0.0001). The results of this study suggest that these proteins play important roles in pulmonary inflammatory reactions elicited against etiological viral agents. In addition, these mediators may affect the process of lung remodeling and the development of pulmonary fibrosis.