Merck
  • MicroRNA-based conversion of human fibroblasts into striatal medium spiny neurons.
MicroRNA-based conversion of human fibroblasts into striatal medium spiny neurons.
Nature protocols (2015-09-18)
Michelle Richner, Matheus B Victor, Yangjian Liu, Daniel Abernathy, Andrew S Yoo
ABSTRACT
The ability to generate human neurons of specific subtypes of clinical importance offers experimental platforms that may be instrumental for disease modeling. We recently published a study demonstrating the use of neuronal microRNAs (miRNAs) and transcription factors to directly convert human fibroblasts to a highly enriched population of striatal medium spiny neurons (MSNs), a neuronal subpopulation that has a crucial role in motor control and harbors selective susceptibility to cell death in Huntington's disease. Here we describe a stepwise protocol for the generation of MSNs by direct neuronal conversion of human fibroblasts in 30 d. We provide descriptions of cellular behaviors during reprogramming and crucial steps involved in gene delivery, cell adhesion and culturing conditions that promote cell survival. Our protocol offers a unique approach to combine microRNAs and transcription factors to guide the neuronal conversion of human fibroblasts toward a specific neuronal subtype.
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-GABA antibody produced in mouse, clone GB-69, ascites fluid
Sigma-Aldrich
Valproic Acid, Sodium Salt, A cell-permeable, short-chained fatty acid that inhibits histone deacetylase (IC₅₀ = 400 µM for HDAC1).
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Alconox® detergent, 0.5 oz packs
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution