Merck
  • The in vitro and in vivo profile of aclidinium bromide in comparison with glycopyrronium bromide.
The in vitro and in vivo profile of aclidinium bromide in comparison with glycopyrronium bromide.
Pulmonary pharmacology & therapeutics (2014-06-15)
Amadeu Gavaldà, Israel Ramos, Carla Carcasona, Elena Calama, Raquel Otal, José Luis Montero, Sonia Sentellas, Monica Aparici, Dolors Vilella, Joan Alberti, Jorge Beleta, Montserrat Miralpeix
ABSTRACT
This study characterised the in vitro and in vivo profiles of two novel long-acting muscarinic antagonists, aclidinium bromide and glycopyrronium bromide, using tiotropium bromide and ipratropium bromide as comparators. All four antagonists had high affinity for the five muscarinic receptor sub-types (M1-M5); aclidinium had comparable affinity to tiotropium but higher affinity than glycopyrronium and ipratropium for all receptors. Glycopyrronium dissociated faster from recombinant M3 receptors than aclidinium and tiotropium but more slowly than ipratropium; all four compounds dissociated more rapidly from M2 receptors than from M3 receptors. In vitro, aclidinium, glycopyrronium and tiotropium had a long duration of action at native M3 receptors (>8 h versus 42 min for ipratropium). In vivo, all compounds were equi-potent at reversing acetylcholine-induced bronchoconstriction. Aclidinium, glycopyrronium and ipratropium had a faster onset of bronchodilator action than tiotropium. Aclidinium had a longer duration of action than glycopyronnium (time to 50% recovery of effect [t½ offset] = 29 h and 13 h, respectively); these compare with a t½ offset of 64 h and 8 h for tiotropium and ipratropium, respectively. Aclidinium was less potent than glycopyrronium and tiotropium at inhibiting salivation in conscious rats (dose required to produce half-maximal effect [ED50] = 38, 0.74 and 0.88 μg/kg, respectively) and was more rapidly hydrolysed in rat, guinea pig and human plasma compared with glycopyrronium or tiotropium. These results indicate that while aclidinium and glycopyrronium are both potent antagonists at muscarinic receptors with similar kinetic selectivity for M3 receptors versus M2, aclidinium has a longer dissociation half-life at M3 receptors and a longer duration of bronchodilator action in vivo than glycopyrronium. The rapid plasma hydrolysis of aclidinium, coupled to its kinetic selectivity, may confer a reduced propensity for systemic anticholinergic side effects with aclidinium versus glycopyrronium and tiotropium.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Propofol, European Pharmacopoeia (EP) Reference Standard
Carbachol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC