Merck
  • Cholesterol-lowering effect of rice bran protein containing bile acid-binding proteins.
Cholesterol-lowering effect of rice bran protein containing bile acid-binding proteins.
Bioscience, biotechnology, and biochemistry (2014-11-07)
Jilite Wang, Masaya Shimada, Yukina Kato, Mio Kusada, Satoshi Nagaoka
ABSTRACT
Dietary plant protein is well known to reduce serum cholesterol levels. Rice bran is a by-product of rice milling and is a good source of protein. The present study examined whether feeding rats a high-cholesterol diet containing 10% rice bran protein (RBP) for 10 d affected cholesterol metabolism. Rats fed dietary RBP had lower serum total cholesterol levels and increased excretion of fecal steroids, such as cholesterol and bile acids, than those fed dietary casein. In vitro assays showed that RBP strongly bound to taurocholate, and inhibited the micellar solubility of cholesterol, compared with casein. Moreover, the bile acid-binding proteins of the RBP were eluted by a chromatographic column conjugated with cholic acid, and one of them was identified as hypothetical protein OsJ_13801 (NCBI accession No. EAZ29742) using MALDI-TOF mass spectrometry analysis. These results suggest that the hypocholesterolemic action of the RBP may be caused by the bile acid-binding proteins.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Sodium cholate hydrate, ≥97.0% (dried material, NT)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Sodium cholate hydrate, BioXtra, ≥99%
Sigma-Aldrich
Sodium cholate hydrate, from bovine and/or ovine bile, ≥99%
Sigma-Aldrich
Sodium cholate hydrate, BioReagent, suitable for cell culture
Supelco
Oleic acid, Selectophore, ≥99%
USP
Glyceryl monooleate, 90%, United States Pharmacopeia (USP) Reference Standard
SAFC
Sodium cholate hydrate
Oleic acid, European Pharmacopoeia (EP) Reference Standard