Merck
  • Preparation of a stir bar coated with molecularly imprinted polymer and its application in analysis of dopamine in urine.
Preparation of a stir bar coated with molecularly imprinted polymer and its application in analysis of dopamine in urine.
Journal of pharmaceutical and biomedical analysis (2014-02-25)
Yun Lei, Guanhong Xu, Fangdi Wei, Jing Yang, Qin Hu
ABSTRACT
A molecularly imprinted polymer coated stir bar (MIP-SB) using dopamine as a template was fabricated. The adsorptive capacity of the MIP-SB was almost 4 times over that of non-imprinted stir bar. The MIP-SB showed good extracting selectivity for dopamine when used to adsorb dopamine and its analogs. An analytical method to determine dopamine in urine sample was established by combining MIP-SB sorptive extraction with HPLC-fluorescence detector. The linear range of dopamine concentration was 0.378-18.9ng/ml with correlation coefficient of 0.9990, and the limit of quantification was about 0.0945ng/ml (S/N=10). The recoveries of dopamine with spiked urine samples at three different levels were between 92.3 and 106.9%, and the relative standard deviations were within 13.2% (n=3). The method is simple and suitable for the determination of dopamine in human urine for clinical application.
Product Description
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
Propyl methacrylate, contains ~200 ppm MEHQ, 97%
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Adrenaline with impurity F, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acrylamide, ≥98.0%
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland