Merck
  • Formulation, characterization and optimization of liposomes containing eicosapentaenoic and docosahexaenoic acids; a methodology approach.
Formulation, characterization and optimization of liposomes containing eicosapentaenoic and docosahexaenoic acids; a methodology approach.
Iranian journal of pharmaceutical research : IJPR (2014-09-23)
Zahra Hadian, Mohammad Ali Sahari, Hamid Reza Moghimi, Mohsen Barzegar
ABSTRACT
Omega-3 fatty acids (FAs) have been shown to prevent cardiovascular disease. The most commonly used omega-3 fatty acids like eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) are highly vulnerable to oxidation and therefore, have short shelf life. Recent advances in nanoliposomes provided a biocompatible system for stabilizing omega-3 FAs. Several methods could be implemented to prepare nanoliposomes. To the best of our knowledge, the performances of these methods in preparation omega-3 FAs have not been examined. Nanoliposomes were prepared by thin film hydration followed by one of the following methods: 1- extrusion, ultrasonic irradiation; 2- bath sonication; 3- probe sonication; or 4- combined probe and bath sonication. The size of liposomes obtained from methods 1 to 4 were 99.7 ± 3.5, 381.2 ± 7.8, 90.1 ± 2.3, and 87.1 ± 4.10 nm with zeta potential being -42.4 ± 1.7, -36.3 ± 1.6, -43.8 ± 2.4, and 31.6 ± 1.9 mV, respectively. The encapsulation efficiency (EE) for DHA was 13.2 ± 1.1%, 26.7 ± 1.9%, 56.9 ± 5.2% and 51.8 ± 3.8% for methods 1 to 4, respectively. The corresponding levels for EPA were 6.5 ± 1.3%, 18.1 ± 2.3%, 38.6 ± 1.8%, and 38 ± 3.7%, respectively. The EE for DHA and EPA of liposomes for both methods 3 and 4 increased significantly (p<0.05). Propanal, as the major volatile product formed during liposomal preparations, amounts from 81.2 ± 4.1 to 118.8 ± 2.3 μg/Kg. The differential scanning calorimetry (DSC) study showed that DHA and EPA influence the phase transition temperature of small unilamellar vesicles (SUVs) of dipalmitoyl phosphatidyl choline (DPPC). Transmission electron microscopy (TEM) images of liposomes stained with uranyl acetate showed that the liposomes were spherical in shape and maintain high structural integrity. In conclusion, probe ultrasound of pre-formed liposomes facilitates significant loading of DHA and EPA into the nanoliposomal membrane.
Sigma-Aldrich
Pentadecanoic acid, 99%
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Pentadecanoic acid, ~99% (capillary GC)
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Hexanal, ≥97%, FCC, FG
Supelco
Pentadecanoic acid, analytical standard
Supelco
Dodecane, analytical standard
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Hexanal, analytical standard
Sigma-Aldrich
Dodecane, ReagentPlus®, ≥99%
Supelco
Valeraldehyde, analytical standard
Sigma-Aldrich
Valeraldehyde, ≥97%, FG
Sigma-Aldrich
Valeraldehyde, 97%
Sigma-Aldrich
Heptaldehyde, 95%
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Supelco
Heptaldehyde, analytical standard
Supelco
Propionaldehyde, analytical standard
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Propionaldehyde, reagent grade, 97%