Merck
  • Lipid-based capsaicin-loaded nano-colloidal biocompatible topical carriers with enhanced analgesic potential and decreased dermal irritation.
Lipid-based capsaicin-loaded nano-colloidal biocompatible topical carriers with enhanced analgesic potential and decreased dermal irritation.
Journal of liposome research (2014-04-29)
Kaisar Raza, Mohammad Afroz Shareef, Parteek Singal, Gajanand Sharma, Poonam Negi, Om Prakash Katare
ABSTRACT
Capsaicin (CP), a recent FDA-approved drug for the topical treatment of neuropathic pain, is associated with several side effects like irritation, burning sensation, and erythema, resulting in poor patient compliance. The present study is an attempt to study the effect of CP encasement in nano-lipoidal carriers (NLCs) on skin-transport characteristics, in vivo pharmacological performance, skin compliance, and stability of the finished product. The study also compares two methods of NLC preparation, namely microemulsification and rotary-evaporation for various attributes. The results demonstrated that microemulsion technique produced smaller nanoparticles vis-à-vis the rotary-evaporation method. Out of the various studied solid lipids, NLCs from stearic acid offered smallest size and the highest negative zeta potential. The NLC-gel offered higher skin permeation and skin retention of CP across LACA mice skin as compared with the conventional cream. The analgesic effect was observed to be enhanced substantially than that of the conventional cream when tested on a radiant mouse tail-flick model. The most alarming problems of skin-irritation and redness were successfully taken care by NLC-gel while the mice group receiving conventional cream showed marked changes in the skin histopathology. Besides the enhanced efficacy and decreased skin-irritation, the developed CP-NLCs also found to be stable and rheologically accepted formulation for the treatment of pain-associated disorders.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Cetyl Palmitate, Pharmaceutical Secondary Standard; Certified Reference Material
Cetyl palmitate 15, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Palmityl palmitate, ≥99%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)