Merck
  • Breath biomarkers of whole-body gamma irradiation in the Göttingen minipig.
Breath biomarkers of whole-body gamma irradiation in the Göttingen minipig.
Health physics (2015-03-27)
Michael Phillips, Renee N Cataneo, Anirudh Chaturvedi, Peter D Kaplan, Mark Libardoni, Mayur Mundada, Urvish Patel, Karla D Thrall, Xiang Zhang
ABSTRACT
There is widespread interest in the development of tools to estimate radiation exposures. Exhaled breath provides a novel matrix for assessing biomarkers that could be correlated with exposures. The use of exhaled breath for estimating radiation exposure is warranted, as studies have shown that external exposure to ionizing radiation causes oxidative stress that accelerates lipid peroxidation of polyunsaturated fatty acids, liberating alkanes and alkane metabolites that are excreted in the breath as volatile organic compounds (VOCs). As a proof of principle study, small groups (n = 4) of Göttingen minipigs were whole-body irradiated with gamma rays delivered by a 60Co source at absorbed doses of 0, 0.25, 0.5, 0.75, 1, 1.25, 2, and 4 Gy. Additional groups (n = 4) were treated with lipopolysaccharide (LPS) or granulocyte colony stimulating factor (G-CSF), with and without concurrent 60Co exposure, at an absorbed dose of 1 Gy. Breath and background air VOC samples were collected on days -3, -2, -1, 0 pre-irradiation, then at 0.25, 24, 48, 72, and 168 h post-irradiation. VOCs were analyzed by automated thermal desorption with two-dimensional gas chromatography and time-of-flight mass spectrometry (ATD GCxGC TOF MS). The results show significant changes in 58 breath VOCs post-irradiation, mainly consisting of methylated and other derivatives of alkanes, alkenes, and benzene. Using a multivariate combination of these VOCs, a radiation response function was constructed, which was significantly elevated at 15 min post irradiation and remained elevated throughout the study (to 168 h post irradiation). As a binary test of radiation absorbed doses ≥ 0.25 Gy, the radiation response function distinguished irradiated animals from shams (0 Gy) with 83-84% accuracy. A randomly derived radiation response function was robust: When half of the biomarkers were removed, accuracy was 75%. An optimally derived function with two biomarkers was 82% accurate. As a binary test of radiation absorbed doses ≥ 0.5 Gy, the radiation response function identified irradiated animals with an accuracy of 87% at 15 min post irradiation and 75.5% at 168 h post irradiation. Treatment with LPS and G-CSF did not affect the radiation response function. This proof-of-principle study supports the hypothesis that breath VOCs may be used for estimating radiation exposures. Further studies will be required to validate the sensitivity and specificity of these potential biomarkers.
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Supelco
4-Bromofluorobenzene, analytical standard
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Methane, electronic grade, ≥99.998%
Sigma-Aldrich
1-Bromo-4-fluorobenzene, 99%
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
5-Norbornene-2-endo,3-exo-dicarboxylic acid, 97%
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon