Merck
  • Effects of type 1 diabetes, sprint training and sex on skeletal muscle sarcoplasmic reticulum Ca2+ uptake and Ca2+-ATPase activity.
Effects of type 1 diabetes, sprint training and sex on skeletal muscle sarcoplasmic reticulum Ca2+ uptake and Ca2+-ATPase activity.
The Journal of physiology (2013-12-04)
A R Harmer, P A Ruell, S K Hunter, M J McKenna, J M Thom, D J Chisholm, J R Flack
ABSTRACT
Calcium cycling is integral to muscle performance during the rapid muscle contraction and relaxation of high-intensity exercise. Ca(2+) handling is altered by diabetes mellitus, but has not previously been investigated in human skeletal muscle. We investigated effects of high-intensity exercise and sprint training on skeletal muscle Ca(2+) regulation among men and women with type 1 diabetes (T1D, n = 8, 3F, 5M) and matched non-diabetic controls (CON, n = 8, 3F, 5M). Secondarily, we examined sex differences in Ca(2+) regulation. Subjects undertook 7 weeks of three times-weekly cycle sprint training. Before and after training, performance was measured, and blood and muscle were sampled at rest and after high-intensity exercise. In T1D, higher Ca(2+)-ATPase activity (+28%) and Ca(2+) uptake (+21%) than in CON were evident across both times and days (P < 0.05), but performance was similar. In T1D, resting Ca(2+)-ATPase activity correlated with work performed until exhaustion (r = 0.7, P < 0.01). Ca(2+)-ATPase activity, but not Ca(2+) uptake, was lower (-24%, P < 0.05) among the women across both times and days. Intense exercise did not alter Ca(2+)-ATPase activity in T1D or CON. However, sex differences were evident: Ca(2+)-ATPase was reduced with exercise among men but increased among women across both days (time × sex interaction, P < 0.05). Sprint training reduced Ca(2+)-ATPase (-8%, P < 0.05), but not Ca(2+) uptake, in T1D and CON. In summary, skeletal muscle Ca(2+) resequestration capacity was increased in T1D, but performance was not greater than CON. Sprint training reduced Ca(2+)-ATPase in T1D and CON. Sex differences in Ca(2+)-ATPase activity were evident and may be linked with fibre type proportion differences.
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard