Merck
  • Histidine(7.36(305)) in the conserved peptide receptor activation domain of the gonadotropin releasing hormone receptor couples peptide binding and receptor activation.
Histidine(7.36(305)) in the conserved peptide receptor activation domain of the gonadotropin releasing hormone receptor couples peptide binding and receptor activation.
Molecular and cellular endocrinology (2015-01-15)
Nkateko M I Mayevu, Han Choe, Ruben Abagyan, Jae Young Seong, Robert P Millar, Arieh A Katz, Colleen A Flanagan
ABSTRACT
Transmembrane helix seven residues of G protein-coupled receptors (GPCRs) couple agonist binding to a conserved receptor activation mechanism. Amino-terminal residues of the GnRH peptide determine agonist activity. We investigated GnRH interactions with the His(7.36(305)) residue of the GnRH receptor, using functional and computational analysis of modified GnRH receptors and peptides. Non-polar His(7.36(305)) substitutions decreased receptor affinity for GnRH four- to forty-fold, whereas GnRH signaling potency was more decreased (~150-fold). Uncharged polar His(7.36(305)) substitutions decreased GnRH potency, but not affinity. [2-Nal(3)]-GnRH retained high affinity at receptors with non-polar His(7.36(305)) substitutions, supporting a role for His(7.36(305)) in recognizing Trp(3) of GnRH. Compared with GnRH, [2-Nal(3)]-GnRH potency was lower at the wild type GnRH receptor, but unchanged or higher at mutant receptors. Results suggest that His(7.36(305)) of the GnRH receptor forms two distinct interactions that determine binding to Trp(3) and couple agonist binding to the conserved transmembrane domain network that activates GPCRs.
Product Description
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%