Merck
  • Liquid crystalline phase as a probe for crystal engineering of lactose: carrier for pulmonary drug delivery.
Liquid crystalline phase as a probe for crystal engineering of lactose: carrier for pulmonary drug delivery.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-12-03)
Sharvil S Patil, Kakasaheb R Mahadik, Anant R Paradkar
ABSTRACT
The current work was undertaken to assess suitability of liquid crystalline phase for engineering of lactose crystals and their utility as a carrier in dry powder inhalation formulations. Saturated lactose solution was poured in molten glyceryl monooleate which subsequently transformed into gel. The gel microstructure was analyzed by PPL microscopy and SAXS. Lactose particles recovered from gels after 48 h were analyzed for polymorphism using techniques such as FTIR, XRD, DSC and TGA. Particle size, morphology and aerosolisation properties of prepared lactose were analyzed using Anderson cascade impactor. In situ seeding followed by growth of lactose crystals took place in gels with cubic microstructure as revealed by PPL microscopy and SAXS. Elongated (size ∼ 71 μm) lactose particles with smooth surface containing mixture of α and β-lactose was recovered from gel, however percentage of α-lactose was more as compared to β-lactose. The aerosolisation parameters such as RD, ED, %FPF and % recovery of lactose recovered from gel (LPL) were found to be comparable to Respitose® ML001. Thus LC phase (cubic) can be used for engineering of lactose crystals so as to obtain particles with smooth surface, high elongation ratio and further they can be used as carrier in DPI formulations.
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Lactose monohydrate, SAJ special grade
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salbutamol hemisulfate salt, ≥98%
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
USP
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
USP
Glyceryl monooleate, 90%, United States Pharmacopeia (USP) Reference Standard