Merck
  • Characterization of antibiotic and disinfectant susceptibility profiles among Pseudomonas aeruginosa veterinary isolates recovered during 1994-2003.
Characterization of antibiotic and disinfectant susceptibility profiles among Pseudomonas aeruginosa veterinary isolates recovered during 1994-2003.
Journal of applied microbiology (2014-11-29)
R C Beier, S L Foley, M K Davidson, D G White, P F McDermott, S Bodeis-Jones, S Zhao, K Andrews, T L Crippen, C L Sheffield, T L Poole, R C Anderson, D J Nisbet
ABSTRACT
To evaluate susceptibility of Pseudomonas aeruginosa veterinary isolates to antibiotics and disinfectants. Pseudomonas aeruginosa isolates collected from dogs (n = 155) and other animals (n = 20) from sixteen states during 1994-2003 were tested for susceptibility. Most isolates were resistant to twenty-one antimicrobials tested, and the highest prevalence of resistance was to β-lactams (93.8%) and sulphonamides (93.5%). Fluoroquinolone resistance did not increase from 1994 to 2003. Ciprofloxacin and enrofloxacin had a 5 and 16% prevalence of resistance, respectively, while sarafloxacin and nalidixic acid had a prevalence of resistance of 97 and 98%, respectively. Strains were pan-resistant to triclosan and chlorhexidine, were highly resistant to benzalkonium chloride and demonstrated high susceptibility to other disinfectants. Didecyldimethylammonium chloride was the most active ammonium chloride. Inducible resistance was observed to cetyl ammonium halides, chlorhexidine and benzyl ammonium chlorides, which formulate disinfectants used in veterinary clinics and dairies. Organic acid inhibition was associated with the dissociated acid species. Dissociated organic acids appear able to inhibit Ps. aeruginosa, and rates of fluoroquinolone resistance merit sustained companion animal isolate surveillance. This is the first report of Ps. aeruginosa susceptibility to 24 disinfectants and illustrates the high resistance of Ps. aeruginosa to both antibiotics and disinfectants.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Benzyldimethylhexadecylammonium chloride, cationic detergent
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Chlorhexidine diacetate salt hydrate, bis(biguanide) antimicrobial
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)