Merck
  • Obeticholic acid, a farnesoid X receptor agonist, improves portal hypertension by two distinct pathways in cirrhotic rats.
Obeticholic acid, a farnesoid X receptor agonist, improves portal hypertension by two distinct pathways in cirrhotic rats.
Hepatology (Baltimore, Md.) (2013-11-22)
Len Verbeke, Ricard Farre, Jonel Trebicka, Mina Komuta, Tania Roskams, Sabine Klein, Ingrid Vander Elst, Petra Windmolders, Tim Vanuytsel, Frederik Nevens, Wim Laleman
ABSTRACT
The farnesoid X receptor (FXR) is a nuclear bile acid receptor involved in bile acid homeostasis, hepatic and intestinal inflammation, liver fibrosis, and cardiovascular disease. We studied the effect of short-term treatment with obeticholic acid (INT-747), a potent selective FXR agonist, on intrahepatic hemodynamic dysfunction and signaling pathways in different rat models of cirrhotic portal hypertension (PHT). For this, thioacetamide (TAA)-intoxicated and bile-duct-ligated (BDL) rats were used as models. After gavage of two doses of 30 mg/kg of INT-747 or vehicle within 24 hours, in vivo hemodynamics were assessed. Additionally, we evaluated the direct effect of INT-747 on total intrahepatic vascular resistance (IHVR) and intrahepatic vascular tone (endothelial dysfunction and hyperresponsiveness to methoxamine) by means of an in situ liver perfusion system and on hepatic stellate cell contraction in vitro. FXR expression and involved intrahepatic vasoactive pathways (e.g., endothelial nitric oxide synthase [eNOS], Rho-kinase, and dimethylarginine dimethylaminohydrolase [DDAH]) were analyzed by immunohistochemistry, reverse-transcriptase polymerase chain reaction, or western blotting. In both cirrhotic models, FXR expression was decreased. Treatment with INT-747 in TAA and BDL reactivated the FXR downstream signaling pathway and decreased portal pressure by lowering total IHVR without deleterious systemic hypotension. In the perfused TAA and BDL cirrhotic liver, INT-747 improved endothelial vasorelaxation capacity, but not hyperresponsiveness. In both groups, this was associated with an increased eNOS activity, which, in TAA, related to down-regulation of Rho-kinase and in BDL to up-regulation of DDAH-2. FXR agonist INT-747 improves PHT in two different rat models of cirrhosis by decreasing IHVR. This hemodynamic effect relates to increased intrahepatic eNOS activity by pathways that differ depending on the etiology of cirrhosis.
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.8%
Sigma-Aldrich
Acetone, ≥99.5%
Sigma-Aldrich
Acetone, JIS 5000
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Anti-phospho-VASP (Ser239) Antibody, clone 16C2, clone 16C2, Upstate®, from mouse
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride