Merck
  • Evaluation of Chlorpheniramine Maleate microparticles in orally disintegrating film and orally disintegrating tablet for pediatrics.
Evaluation of Chlorpheniramine Maleate microparticles in orally disintegrating film and orally disintegrating tablet for pediatrics.
Drug development and industrial pharmacy (2013-04-30)
Hao Lou, Min Liu, Wen Qu, Zheyi Hu, Ed Brunson, James Johnson, Hassan Almoazen
ABSTRACT
To mask the bitterness of Chlorpheniramine Maleate via encapsulating drug into Eudragit EPO microparticles, and then incorporate these microparticles into orally disintegrating films (ODF) and orally disintegrating tablets (ODT) for pediatric uses. Spray drying of water-in-oil emulsion was utilized to encapsulate Chlorpheniramine Maleate into Eudragit EPO microparticles. Based on an orthogonal experimental design L9 (3(3)), polynomial regression models were developed to evaluate correlation between microparticle properties (encapsulation efficiency and drug release) and variables (X1: weight ratio of polymer to drug, X2: volume ratio of oil to water and X3: Q-flow of spray dryer). ODF and ODT formulations were evaluated including weight variation, content uniformity, tensile strength, disintegration time, friability and dissolution profiles. The bitterness taste test was evaluated in 10 adult volunteers. From polynomial regression analysis, the best values of variables leading to the optimized microparticles were X1 = 10, X2 = 3 and X3 = 45. The optimized microparticles were incorporated into ODF and ODT with satisfactory weight and drug content uniformity, and acceptable physical strength. Both dosage forms disintegrated immediately (less than 40 s) in simulated saliva solutions. The outcome of taste-masking test indicated that microparticles alleviated drug bitterness significantly; bitterness was not discernible with microparticles incorporated in ODT, whereas only slight bitterness was detected from microparticles incorporated into ODF. Both ODF and ODT are shown to be suitable vehicles for taste masked Chlorpheniramine Maleate microparticles with potential for pediatric uses.
Product Description
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Chlorpheniramine maleate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)