Merck
  • Inhibin alpha-subunit (INHA) expression in adrenocortical cancer is linked to genetic and epigenetic INHA promoter variation.
Inhibin alpha-subunit (INHA) expression in adrenocortical cancer is linked to genetic and epigenetic INHA promoter variation.
PloS one (2014-08-12)
Johannes Hofland, Jacobie Steenbergen, Jacoba M Voorsluijs, Michael M P J Verbiest, Ronald R de Krijger, Leo J Hofland, Wouter W de Herder, Andre G Uitterlinden, Richard A Feelders, Frank H de Jong
ABSTRACT
Adrenocortical carcinoma (ACC) is a rare, but highly malignant tumor of unknown origin. Inhibin α-subunit (Inha) knockout mice develop ACCs following gonadectomy. In man, INHA expression varies widely within ACC tissues and its circulating peptide inhibin pro-αC has been described as a novel tumor marker for ACC. We investigated whether genetic and epigenetic changes of the INHA gene in human ACC cause loss or variation of INHA expression. To this end, analyses of INHA sequence, promoter methylation and mRNA expression were performed in human adrenocortical tissues. Serum inhibin pro-αC levels were also measured in ACC patients. INHA genetic analysis in 37 unique ACCs revealed 10 novel, heterozygous rare variants. Of the 3 coding bases affected, one variant was synonymous and two were missense variants: S72F and S184F. The minor allele of rs11893842 at -124 bp was observed at a low frequency (24%) in ACC samples and was associated with decreased INHA mRNA levels: 4.7±1.9 arbitrary units for AA, compared to 26±11 for AG/GG genotypes (P = 0.034). The methylation of four proximal INHA promoter CpGs was aberrantly increased in five ACCs (47.7±3.9%), compared to normal adrenals (18.4±0.6%, P = 0.0052), whereas the other 14 ACCs studied showed diminished promoter methylation (9.8±1.1%, P = 0.020). CpG methylation was inversely correlated to INHA mRNA levels in ACCs (r = -0.701, p = 0.0036), but not associated with serum inhibin pro-αC levels. In conclusion, aberrant methylation and common genetic variation in the INHA promoter occur in human ACCs and are associated with decreased INHA expression.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%