Merck
  • The antipsychotic amisulpride: ultrastructural evidence of its secretory activity in salivary glands.
The antipsychotic amisulpride: ultrastructural evidence of its secretory activity in salivary glands.
Oral diseases (2013-11-20)
F Loy, M Isola, R Isola, M A Lilliu, P Solinas, G Conti, T Godoy, A Riva, J Ekström
ABSTRACT
Amisulpride is reported to inhibit clozapine-induced sialorrhea. Preclinically, clozapine evokes muscarinic-M1-type-mediated secretion that, however, amisulpride does not reduce. Instead, amisulpride, without causing any overt secretion per se, enhances both nerve- and autonomimetic-evoked salivation by unknown mechanism(s). Hypothesizing that amisulpride prepares the gland for secretion, we looked for ultrastructural events indicating secretory activity in intercellular canaliculi of serous/seromucous cells, that is, density increase in protrusions (reflecting anchored granules) and in microbuds (reflecting recycling membranes and/or vesicle secretion) and decrease in microvilli (reflecting the cytoskeletal re-arrangement related to exocytosis). Rat parotid and submandibular glands were exposed to amisulpride in vivo or in vitro. Glands were processed for transmission electron and scanning electron microscopy and then morphometrically assessed. Cells were packed with secretory granules. The density of protrusions increased in both glands, whereas significant and parallel changes in microvilli and microbuds occurred only in parotid glands, and in vitro. Amisulpride induced ultrastructural signs of secretory activity but to varying extent; in submandibular glands, in contrast to parotid glands, changes were not brought beyond the granular anchoring stage. Amisulpride may provide an overall readiness for secretion that will result in augmented responses to agonists, a phenomenon of potential interest in dry-mouth treatment.
Sigma-Aldrich
Amisulpride, ≥98% (HPLC)
Amisulpride, European Pharmacopoeia (EP) Reference Standard
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
USP
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Supelco
(±)-Propranolol hydrochloride, analytical standard
Supelco
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Amisulpride for system suitability, European Pharmacopoeia (EP) Reference Standard
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard