Merck
  • Cimetidine attenuates vinorelbine-induced phlebitis in mice by militating E-selectin expression.
Cimetidine attenuates vinorelbine-induced phlebitis in mice by militating E-selectin expression.
Cancer chemotherapy and pharmacology (2014-06-01)
Zhuo Wang, Lijuan Ma, Xuebin Wang, Heping Cai, Jin Huang, Jiyong Liu, Jinhong Hu, Dingfeng Su
ABSTRACT
We investigated E-selectin expression in mice and rabbits with vinorelbine-induced phlebitis and the effect of cimetidine. To find the relationship between E-selectin expression and vinorelbine-induced phlebitis. Mouse and rabbit model of vinorelbine-induced phlebitis was established by intravenous infusion of vinorelbine. Pathological observation, molecular-biological determination of E-selectin and protein function of it was evaluated. Grossly, we observed swelling, edema and cord-like vessel changes in mice receiving vinorelbine but only mild edema in mice pretreated with cimetidine. Pathological scoring yielded a total score of 37 for vinorelbine-treated mice and 17 for mice pretreated with cimetidine (P < 0.05). ELISA revealed that rabbits treated with vinorelbine had markedly higher serum contents of E-selectin than normal saline (NS) controls (vinorelbine 1.534 ± 0.449 vs. NS 0.746 ± 0.170 ng/mL, P < 0.05), which was markedly attenuated by cimetidine (cimetidine 0.717 ± 0.468 vs. vinorelbine 1.534 ± 0.449 ng/mL, P < 0.05). Rose Bengal staining assays showed that vinorelbine markedly increased the adhesion rate of neutrophils for endothelial cells (vinorelbine 38.70 ± 8.34% vs. controls 8.93 ± 4.85%, P < 0.01), which, however, was significantly suppressed by cimetidine (9.93 ± 5.91%, P < 0.01 vs. vinorelbine). In E-selectin knockout mice, we found no apparent difference in tail swelling in mice receiving vinorelbine or cimetidine and vinorelbine. In conclusion, cimetidine attenuates vinorelbine-induced phlebitis in mice probably by suppressing increased expression of E-selectin.
Product Description
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cimetidine
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cimetidine, United States Pharmacopeia (USP) Reference Standard
Cimetidine, European Pharmacopoeia (EP) Reference Standard