Merck
  • Leptin administration activates irisin-induced myogenesis via nitric oxide-dependent mechanisms, but reduces its effect on subcutaneous fat browning in mice.
Leptin administration activates irisin-induced myogenesis via nitric oxide-dependent mechanisms, but reduces its effect on subcutaneous fat browning in mice.
International journal of obesity (2005) (2014-09-10)
A Rodríguez, S Becerril, L Méndez-Giménez, B Ramírez, N Sáinz, V Catalán, J Gómez-Ambrosi, G Frühbeck
ABSTRACT
BACKGROUND/OBJETIVES: Obese leptin-deficient ob/ob mice exhibit high adiposity and reduced muscle mass with leptin replacement promoting weight loss and inducing muscle accretion through PGC-1α-dependent mechanisms. Our aim was to analyze in vivo and in vitro the effect of leptin on FNDC5, a novel PGC-1α-dependent myokine that is synthesized and cleaved to form irisin that induces white adipose browning. Twelve-week-old male wild-type and ob/ob mice were divided in three groups as follows: control, leptin-treated (1 mg kg(-1) day(-1)) and pair-fed. Leptin administration was associated with increased gastrocnemius weight and cell surface area, higher Pgc1a and Fndc5 transcript levels and a slight increase in circulating irisin. Leptin upregulated Fndc5 expression through nitric oxide (NO)-dependent mechanisms in murine C2C12 myocytes and stimulated both basal and irisin-stimulated myogenesis, as evidenced by increased myocyte cell proliferation, higher myogenin and myonectin transcript levels together with lower mRNA expression of myostatin and dystrophin and the muscle atrophy-related factors MuRF1 and MAFbx. Interestingly, leptin downregulated Fndc5 expression in a NO-independent manner in murine differentiated subcutaneous adipocytes. Furthermore, leptin prevented the irisin-induced upregulation of both brown (Ucp1 and Cidec) and beige (Tmem26) adipocyte-specific genes and the increase in uncoupling protein-1-positive cells. Taken together, our results provide evidence for a regulatory role of leptin on FNDC5/irisin, favoring muscle accretion but reducing fat browning.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
L-N6-(1-Iminoethyl)lysine dihydrochloride
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
BrdU Cell Proliferation Assay, This proliferation assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission