Merck
  • Feasibility study on microencapsulation of anaerobic Clostridium acetobutylicum ATCC 824 by emulsification method for application in biobutanol production.
Feasibility study on microencapsulation of anaerobic Clostridium acetobutylicum ATCC 824 by emulsification method for application in biobutanol production.
Journal of microencapsulation (2014-04-05)
Sweta Rathore, Paul Wan Sia Heng, Lai Wah Chan
ABSTRACT
This work evaluated the feasibility of microencapsulating Clostridium acetobutylicum ATCC 824 cells by emulsification for fermentation to produce biobutanol. The effects of selected emulsification process on viability of the vegetative cells and spores were investigated to enable the selection of appropriate form of bacterium for immobilisation. The spores were found to be more suitable for microencapsulation than the vegetative cells. Design of experiment and mathematical models were then used to evaluate the effects of gellan gum concentration, HLB of surfactant blend, temperature and stirring speed on the properties of the microspheres produced. Using the predicted optimal conditions, the spores were successfully immobilised in spherical microspheres for use in fermentation. The microencapsulated spores were easily revived by heat shock treatment and could produce 8.2 g/l of butanol, which was higher than that generally reported in literature. The microencapsulation method developed provides means of producing reusable microbioreactors for anaerobic spore-forming microorganisms.
Sigma-Aldrich
Dextrose monohydrate, 97.5-102.0% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose monohydrate, meets analytical specification of Ph. Eur., BP, Ph Franç., 97.5-102.0% anhydrous basis(HPLC)
Sigma-Aldrich
D-(+)-Glucose monohydrate, tested according to Ph. Eur.
Millipore
D-(+)-Glucose monohydrate, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%