Merck
  • Increased interleukin 21 and follicular helper T-like cells and reduced interleukin 10+ B cells in patients with new-onset systemic lupus erythematosus.
Increased interleukin 21 and follicular helper T-like cells and reduced interleukin 10+ B cells in patients with new-onset systemic lupus erythematosus.
The Journal of rheumatology (2014-07-17)
Li Wang, Pingwei Zhao, Liang Ma, Yuxing Shan, Zhenyu Jiang, Juan Wang, Yanfang Jiang
ABSTRACT
To elucidate the potential role of follicular helper T cells (TFH) and interleukin 10 (IL-10)+ B cells in the development of systemic lupus erythematosus (SLE). The numbers of peripheral blood CD27+, CD38+, CD86+, CD95+, IL-10+ B cells, and inducible T cell costimulator (ICOS)+, programmed death-1 (PD-1)+, IL-21+, CXCR5+CD4+ TFH-like cells were examined in 23 patients with new onset SLE and 20 healthy controls (HC). In comparison with HC, significantly reduced numbers of CD19+ and IL-10+ B cells, but increased numbers of CD27(high), CD86+, CD95+ B cells, CXCR5+CD4+, ICOS+, PD-1+, and IL-21+ TFH-like cells were detected, which were accompanied by higher levels of serum IL-21, but lower levels of IL-10 in the patients. Treatment with anti-SLE therapy modulated the imbalance of different subsets of B and TFH-like cells. The levels of serum IL-21 and IL-10 were positively correlated with the numbers of CD4+CXCR5+ TFH-like and CD19+CD5+CD1d+ B cells in the patients, respectively. The numbers of CD27(high) B cells were correlated positively with IL-21+ TFH-like cells, but negatively with IL-10+ B cells. The values of SLE Disease Activity Index, C3, and erythrocyte sedimentation rate were correlated positively with serum IL-21, but negatively with IL-10 in those patients. Our data indicate that the imbalance of IL-21+ TFH-like, CD27(high), and IL-10+ B cells may be associated with the pathogenesis of SLE, and levels of serum IL-21 and IL-10 may be valuable for evaluating disease activity in SLE.
Product Description
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Prednisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mouse IL-21 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Human IL21 / Interleukin-21 ELISA Kit
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard