Merck
  • A signature predicting poor prognosis in gastric and ovarian cancer represents a coordinated macrophage and stromal response.
A signature predicting poor prognosis in gastric and ovarian cancer represents a coordinated macrophage and stromal response.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-03-25)
Rita A Busuttil, Joshy George, Richard W Tothill, Kylie Ioculano, Adam Kowalczyk, Catherine Mitchell, Stephen Lade, Patrick Tan, Izhak Haviv, Alex Boussioutas
ABSTRACT
Gene-expression profiling has revolutionized the way we think about cancer and confers the ability to observe the synchronous expression of thousands of genes. The use of putative genome-level expression profiles has allowed biologists to observe the complex interactions of genes that constitute recognized biologic pathways. We used gastric and ovarian datasets to identify gene-expression signatures and determine any functional significance. Microarray data of 94-tumor and 45-benign samples derived from patients with gastric cancer were interrogated using Hierarchical Ordered Partitioning and Collapsing Hybrid analysis identifying clusters of coexpressed genes. Clusters were further characterized with respect to biologic significance, gene ontology, and ability to discriminate between normal and tumor tissue. Tumor tissues were separated into epithelial and stromal compartments and immunohistochemical analysis performed to further elucidate specific cell lineages expressing genes contained in the signature. We identified a "stromal-response" expression signature, highly enriched for inflammatory, extracellular matrix, cytokine, and growth factor proteins. The majority of genes in the signature are expressed in the tumor-associated stroma but were absent in associated premalignant conditions. In gastric cancer, this module almost perfectly differentiates tumor from nonmalignant gastric tissue and hence can be regarded as a highly tumor-specific gene-expression signature. We show that these genes are consistently coexpressed across a range of independent gastric datasets as well as other cancer types suggesting a conserved functional role in cancer. In addition, we show that this signature can be a surrogate marker for M2 macrophage activity and has significant prognostic implications in gastric and ovarian high-grade serous cancer.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Phenol, natural, 97%, FG