Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ≥99.5%
Pricing and availability is not currently available.
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri™, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99.0%
Pricing and availability is not currently available.