Merck
Antifungal drugs during pregnancy: an updated review.
The Journal of antimicrobial chemotherapy (2014-09-11)
Benoît Pilmis, Vincent Jullien, Jack Sobel, Marc Lecuit, Olivier Lortholary, Caroline Charlier
ABSTRACT
Antifungal prescription remains a challenge in pregnant women because of uncertainties regarding fetal toxicity and altered maternal pharmacokinetic parameters that may affect efficacy or increase maternal and fetal toxicity. We present updated data reviewing the available knowledge and current recommendations regarding antifungal prescription in pregnancy. Amphotericin B remains the first-choice parenteral drug in spite of its well-established toxicity. Topical drugs are used throughout pregnancy because of limited absorption. Recent data have clarified the teratogenic effect of high-dose fluconazole during the first trimester and provided reassuring cumulative data regarding its use at a single low dose in this key period. Recent data have also provided additional safety data on itraconazole and lipidic derivatives of amphotericin B. Regarding newer antifungal drugs, including posaconazole and echinocandins, clinical data are critically needed before considering prescription in pregnancy.
Product Description
Supelco
Morpholine, analytical standard
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Morpholine, polymer-bound, 200-400 mesh, extent of labeling: 2.75-3.25 mmol/g loading, 1 % cross-linked
Sigma-Aldrich
StratoSpheres PL-MPH resin, 50-100 mesh, extent of labeling: 3.0-4.0 mmol/g loading, 1 % cross-linked
Sigma-Aldrich
Ergosterol, ≥75%
Supelco
Ergosterol, Pharmaceutical Secondary Standard; Certified Reference Material
Ergosterol, European Pharmacopoeia (EP) Reference Standard