Merck
  • Enhancement of extinction learning attenuates ethanol-seeking behavior and alters plasticity in the prefrontal cortex.
Enhancement of extinction learning attenuates ethanol-seeking behavior and alters plasticity in the prefrontal cortex.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-05-30)
Justin T Gass, Heather Trantham-Davidson, Amanda S Kassab, William B Glen, M Foster Olive, L Judson Chandler
ABSTRACT
Addiction is a chronic relapsing disorder in which relapse is often initiated by exposure to drug-related cues. The present study examined the effects of mGluR5 activation on extinction of ethanol-cue-maintained responding, relapse-like behavior, and neuronal plasticity. Rats were trained to self-administer ethanol and then exposed to extinction training during which they were administered either vehicle or the mGluR5 positive allosteric modulator 3-cyano-N-(1,3-diphenyl-1H-pyrazol-5-yl) or CDPPB. CDPPB treatment reduced active lever responding during extinction, decreased the total number of extinction sessions required to meet criteria, and attenuated cue-induced reinstatement of ethanol seeking. CDPPB facilitation of extinction was blocked by the local infusion of the mGluR5 antagonist 3-((2-methyl-4-thiazolyl)ethynyl) pyridine into the infralimbic (IfL) cortex, but had no effect when infused into the prelimbic (PrL) cortex. Analysis of dendritic spines revealed alterations in structural plasticity, whereas electrophysiological recordings demonstrated differential alterations in glutamatergic neurotransmission in the PrL and IfL cortex. Extinction was associated with increased amplitude of evoked synaptic PrL and IfL NMDA currents but reduced amplitude of PrL AMPA currents. Treatment with CDPPB prevented the extinction-induced enhancement of NMDA currents in PrL without affecting NMDA currents in the IfL. Whereas CDPPB treatment did not alter the amplitude of PrL or IfL AMPA currents, it did promote the expression of IfL calcium-permeable GluR2-lacking receptors in both abstinence- and extinction-trained rats, but had no effect in ethanol-naive rats. These results confirm changes in the PrL and IfL cortex in glutamatergic neurotransmission during extinction learning and demonstrate that manipulation of mGluR5 facilitates extinction of ethanol cues in association with neuronal plasticity.
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
CDPPB, ≥98% (HPLC)
Sigma-Aldrich
MTEP hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, JIS special grade