Merck
  • Electrochemical immunosensor for the analysis of the breast cancer biomarker HER2 ECD.
Electrochemical immunosensor for the analysis of the breast cancer biomarker HER2 ECD.
Talanta (2014-08-17)
Raquel C B Marques, Subramanian Viswanathan, Henri P A Nouws, Cristina Delerue-Matos, M Begoña González-García
ABSTRACT
Human epidermal growth factor receptor 2 (HER2) is a breast cancer biomarker that plays a major role in promoting breast cancer cell proliferation and malignant growth. The extracellular domain (ECD) of HER2 can be shed into the blood stream and its concentration is measurable in the serum fraction of blood. In this work an electrochemical immunosensor for the analysis of HER2 ECD in human serum samples was developed. To achieve this goal a screen-printed carbon electrode, modified with gold nanoparticles, was used as transducer surface. A sandwich immunoassay, using two monoclonal antibodies, was employed and the detection of the antibody-antigen interaction was performed through the analysis of an enzymatic reaction product by linear sweep voltammetry. Using the optimized experimental conditions the calibration curve (ip vs. log[HER2 ECD]) was established between 15 and 100 ng/mL and a limit of detection (LOD) of 4.4 ng/mL was achieved. These results indicate that the developed immunosensor could be a promising tool in breast cancer diagnostics, patient follow-up and monitoring of metastatic breast cancer since it allows quantification in a useful concentration range and has an LOD below the established cut-off value (15 ng/mL).
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Magnesium nitrate hexahydrate, ACS reagent, 99%
Sigma-Aldrich
Magnesium nitrate hexahydrate, puriss. p.a., ACS reagent, 98.0-102.0% (KT)
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioXtra, ≥98%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, 0.1 M
Sigma-Aldrich
Nitric acid, 1 M
Sigma-Aldrich
Indoxyl phosphate disodium salt, crystalline
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, SAJ first grade, 65.0-66.0%, density: 1.40
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %